Chemical Containers, Electron Microscopy Sciences
Chemical Containers, Electron Microscopy Sciences
Certificates
Chemical Containers, Electron Microscopy Sciences
Chemical Containers, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Relied on to transport dust samples from the moon, these injection-molded polyethylene containers are ideal for small quantities of liquids or solids.
  • Made of injection-molded polyethylene
  • Screw caps feature a leakproof inner seal and prevent leakage and spill
  • Easy to clean
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Ø ext.
    Availability
    Price
  • 5 ml
    Container
    22 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $135.42
    $135.42
    Each
       $135.42
    Supplier Number: 64299-40
  • 10 ml
    Container
    22 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $158.64
    $158.64
    Each
       $158.64
    Supplier Number: 64299-41
  • 30 ml
    Container
    32 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $162.29
    $162.29
    Each
       $162.29
    Supplier Number: 64299-42
  • 90 ml
    Container
    52 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $193.75
    $193.75
    Each
       $193.75
    Supplier Number: 64299-43
  • 120 ml
    Container
    52 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $239.67
    $239.67
    Each
       $239.67
    Supplier Number: 64299-44
  • 180 ml
    Container
    52 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $248.44
    $248.44
    Each
       $248.44
    Supplier Number: 64299-45