JEOL SEM Mount, Electron Microscopy Sciences | Avantor
JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
Certificates
JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
We offer a complete line of SEM specimen mounts.
  • For scanning electron microscopy
  • Standard lathe finish and economically priced
  • Made from ultra-pure aluminum
Head: 50 mm diameter
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Matière
    Availability
    Price
  • SEM Mount JEOL
    50 x 5 mm
    Aluminum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $144.77
    $144.77
    Each
       $144.77
    Supplier Number: 75705
  • SEM Mount JEOL
    50 x 5 mm
    Aluminum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,281.28
    $1,281.28
    Each
       $1,281.28
    Supplier Number: 75706
  • SEM Mount JEOL
    50 x 10 mm
    Aluminum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $157.88
    $157.88
    Each
       $157.88
    Supplier Number: 75707
  • SEM Mount JEOL
    50 x 10 mm
    Aluminum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,392.25
    $1,392.25
    Each
       $1,392.25
    Supplier Number: 75708