Cylinder Mount Hitachi M4, Electron Microscopy Sciences
Cylinder Mount Hitachi M4, Electron Microscopy Sciences
Cylinder Mount Hitachi M4, Electron Microscopy Sciences
Cylinder Mount Hitachi M4, Electron Microscopy Sciences
Spring-loaded clips hold specimens for scanning electron microscopy without adhesives.
  • Spring loaded clips accommodate samples up to 2 mm thickness
  • Available with single and multiple clips
  • Available in 25, 32, or 50 mm diameter
Samples are held securely by small spring-loaded clips and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time. Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal and more. SEM Clip Mounts with multiple clips can hold larger samples or multiple smaller samples.
Product Family Options
  • Désignation
    Ø
  • SEM Clip, 25 x 6 mm x M4 Cylinder Mount, 1 clip
    25 mm
    Each
    Each - $67.34
    $67.34
    Each
       $67.34
     : 75930-05
  • SEM Clip, 25 x 6 mm x M4 Cylinder Mount, 2 clips
    25 mm
    Each
    Each - $86.50
    $86.50
    Each
       $86.50
     : 75930-10
  • SEM Clip, 25 x 6 mm x M4 Cylinder Mount, 3 clips
    25 mm
    Each
    Each - $103.18
    $103.18
    Each
       $103.18
     : 75930-20
  • SEM Clip, 32 x 10 mm x M4 Cylinder Mount, 1 clip
    32 mm
    Each
    Each - $67.34
    $67.34
    Each
       $67.34
     : 75930-15
  • SEM Clip, 32 x 10 mm x M4 Cylinder Mount, 2 clips
    32 mm
    Each
    Each - $84.47
    $84.47
    Each
       $84.47
     : 75930-25
    Marketsource
    Marketsource
  • SEM Clip, 32 x 10 mm x M4 Cylinder Mount, 3 clips
    32 mm
    Each
    Each - $120.38
    $120.38
    Each
       $120.38
     : 75930-35
    Marketsource
    Marketsource
  • SEM Clip, 50 x 6 mm x M4 Cylinder Mount, 1 clip
    50 mm
    Each
    Each - $91.30
    $91.30
    Each
       $91.30
     : 75954-15
  • SEM Clip, 50 x 6 mm x M4 Cylinder Mount, 2 clips
    50 mm
    Each
    Each - $121.70
    $121.70
    Each
       $121.70
     : 75954-25
  • SEM Clip, 50 x 6 mm x M4 Cylinder Mount, 3 clips
    50 mm
    Each
    Each - $139.87
    $139.87
    Each
       $139.87
     : 75954-30
  • SEM Clip, 50 x 6 mm x M4 Cylinder Mount, 4 clips
    50 mm
    Each
    Each - $170.01
    $170.01
    Each
       $170.01
     : 75954-35
  • SEM Clip, 50 x 6 mm x M4 Cylinder Mount, 8 clips
    50 mm
    Each
    Each - $250.44
    $250.44
    Each
       $250.44
     : 75954-40