For use in Hitachi S-800, S-4000, S-4100, S-4200, S-4300, S-4500, S-4700 and S-3600N SEMs.
- Aluminum or brass with stainless steel allen set screws
- Holds SEM pin mounts
The scanning electron microscope holder is ideal for cross sections up to 6.35 mm (1/4") thick.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.