Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Kit, Electron Microscopy Sciences
Certificates
Brain Punch Kit, Electron Microscopy Sciences
Brain Punch Kit, Electron Microscopy Sciences
9 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique.
Each punch has an electro polished sharp end for smooth cuts and minimal tissue adhesion. The smaller tubes are reinforced by an outer layer of concentric tubing except at the tip.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Brain Punch Handle Only
    Log in for availability
    Log in for availability
    Each
    Each - $475.25
    $475.25
    Each
       $475.25
    Supplier Number: 57400
  • Brain Punch Individual, 0.25 mm
    Discontinued
    Supplier Number: 57391
  • Brain Punch Individual, 0.50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $448.69
    $448.69
    Each
       $448.69
    Supplier Number: 57393
  • Brain Punch Individual, 0.75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $448.69
    $448.69
    Each
       $448.69
    Supplier Number: 57395
  • Brain Punch Individual, 1.0 mm
    Log in for availability
    Log in for availability
    Each
    Each - $448.69
    $448.69
    Each
       $448.69
    Supplier Number: 57397
  • Brain Punch Individual, 1.25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $448.69
    $448.69
    Each
       $448.69
    Supplier Number: 57399
  • Brain Punch Individual, 1.50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $435.36
    $435.36
    Each
       $435.36
    Supplier Number: 57403
  • Brain Punch Individual, 1.75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $435.36
    $435.36
    Each
       $435.36
    Supplier Number: 57404
  • Brain Punch Individual, 2.00 mm
    Log in for availability
    Log in for availability
    Each
    Each - $391.59
    $391.59
    Each
       $391.59
    Supplier Number: 57405
  • Brain Punch Set of Five, 0.25 – 1.25 mm
    Discontinued
    Supplier Number: 57401
  • Brain Punch Set of Three, 1.50 – 2.00 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,605.35
    $1,605.35
    Each
       $1,605.35
    Supplier Number: 57402