HEPES, Free Acid Solutions, ULTROL® Grade, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
Certificates
About this item
A zwitterionic N-substituted amino sulfonic acid buffers active in the pH range of 6.0 to 8.5.
- Absorbance: <0.05 at 260 nm, 1 M, H₂O
- Sterile filtered
- pKa: 7.55 at 20 °C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:7365-45-9
- Formula:C₈H₁₈N₂O₄S
- Formula weight:238.30 g/mol
- Purity:≥99%
- Storage temperature:15 – 30 °C
- Formulation:1.0 M solution
- Physical form:Solution