Separating Funnels, DWK Life Sciences
À propos de cet article
Micro separating funnels manufactured from 33 expansion, low extractable borosilicate glass
Squibb-type funnel with a PTFE stopcock plug with a 2 mm bore.
Certifications: Material conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Series 298280 is supplied with hex-head stopper and series 298281 is supplied without stoppers.
4 Options Available Below
Product Details & Documents
Micro separating funnels manufactured from 33 expansion, low extractable borosilicate glass
Squibb-type funnel with a PTFE stopcock plug with a 2 mm bore.
Certifications: Material conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Series 298280 is supplied with hex-head stopper and series 298281 is supplied without stoppers.
Product Family Options
Informations sur le produit
- DésignationCapacitéHauteurStopper MaterialTop ST Joint SizeAvailabilityPrice
- Supplier Number: 298280-0010
- Supplier Number: 298280-0025
- Micro Separatory Funnel125 ml206 mmPTFE[ST] 14/20Discontinued-Supplier Number: -
- Supplier Number: 298281-0010
- Supplier Number: 298281-0025