Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE, 10X, 1 L, 8.3
Catalog # CAMB-019
Supplier: Rockland Immunochemical
Supplier #: MB-019
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # CAMB-019
Supplier: Rockland Immunochemical
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Désignation:TBE, 10X
- Taille:1 L
- pH:8.3