Bee-Safe® Hood, High-Tech Conversions | Avantor
Bee-Safe® Hood, High-Tech Conversions
Bee-Safe® Hood, High-Tech Conversions
Certificates
Bee-Safe® Hood, High-Tech Conversions
2 Options Available Below
Product Details & Documents
This hood is a disposable, lightweight and economical solution with superior particulate filtration.
  • Spunbound polypropylene nonwoven
  • Disposable
  • Covers hair and neck
  • Lightweight
  • Superior particulate filtration
The Bee-Safe Hood provides hair and neck coverage that is lightweight for comfort and has superior particlate filtration for many different applications
Applications include cleanroom, food industry, medical, pharma, and safety. For use in laboratories and manufacturing environments.
SBPP nonwoven material. Latex-free elastic edges to minimize allergic reactions. Machine made to ensure consistency throughout production. Cleanroom packaging.
This hood is disposable and has very competitive pricing.
Packaging: 14 x 11 x 11"; 1000 Pieces/Case.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76169-758
  • 76169-760
  • Fabric:
  • Spunbound Polypropylene
  • Spunbound Polypropylene
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Weight:
  • 17 g/m²
  • 17 g/m²
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    W×L
    Availability
    Price
  • White Cleanroom Hood
    23×28 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $740.17
    $740.17
    Case of 1,000
       $740.17
    Supplier Number: GAH-HOOD-WH-L
    Spécifications:
    More
    Specifications
    Cat. No.
    76169-758
    Fabric
    Spunbound Polypropylene
    Garment application
    Cleanroom
    Weight
    17 g/m²
  • Blue Cleanroom Hood
    23×28 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,204.96
    $1,204.96
    Case of 1,000
       $1,204.96
    Supplier Number: GAH-HOOD-BL-L
    Spécifications:
    More
    Specifications
    Cat. No.
    76169-760
    Fabric
    Spunbound Polypropylene
    Garment application
    Cleanroom
    Weight
    17 g/m²