About this item
Quick (10 minute) method for separating, cleaning, and concentrating short (< 200 nt) ssDNA or RNA.
- Ideal for non-enzymatic elimination of genomic DNA from transcripts, probes, primers, etc.
- Zymo-Spin™ Column technology allows for elution into minimal volumes (≥ 6 μL).
The ssDNA/RNA Clean & Concentrator™ provides a simple and reliable method for the rapid separation, clean-up, and concentration of up to ~5 μg (per prep.) of single stranded DNA and/ or RNA from double stranded species (e.g. genomic DNA). This simple 10 minutes procedure is based on the use of a unique single-buffer system and Zymo-Spin™ Column technology. Single stranded DNA or RNA ≥ 17 nucleotides (e.g., transcripts, probes, primers) can be safely treated and co-purified using this kit. The highly concentrated, purified DNA/RNA is great for downstream applications including PCR, RT-PCR, hybridization, etc.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:ssDNAssRNA
- Sample type:RNA prepsDNA preps
- Target molecule:Copurification of DNA & RNADNARNA
- Description:ssDNA/RNA Clean & Concentrator™