Eyelash Manipulator; An EM Tool, Electron Microscopy Sciences
Eyelash Manipulator; An EM Tool, Electron Microscopy Sciences
Eyelash Manipulator
Catalog # 103302-486
Supplier #:  71182
Eyelash Manipulator; An EM Tool, Electron Microscopy Sciences
Catalog # 103302-486
Eyelash Manipulator; An EM Tool, Electron Microscopy Sciences
Eyelash Manipulator; An EM Tool, Electron Microscopy Sciences
Eyelash Manipulator
Price
$40.93
Log in for availability
Each Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This is for direct manipulation of sections and small tissue samples.
  • Sable brush handle made from the highest quality wood
  • Cleaned and sterilized with alcohol and a tissue
Suitable for ultramicrotomy to carefully tease sections that are in the diamond knife boat.
Specifications
  • Désignation:
    Eyelash Manipulator