JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
Certificates
JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
JEOL SEM mounts are made from lathe finished ultra-pure aluminum for scanning electron microscopy.
Head diameter: 11/4" (32 mm)
Height: 3/8" (10 mm)
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • SEM Mount JEOL Aluminium 32 mm
    10/pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 89,97 $
    89,97 $
    Each
       89,97 $
    Supplier Number: 75702
  • SEM Mount JEOL Aluminium 32 mm
    50/pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 368,08 $
    368,08 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 315,08 $
    315,08 $
    Each
       368,08 $
    Pack of 50
       315,08 $
    Supplier Number: 75703
    Spécifications:
    Marketsource
    Marketsource
  • SEM Mount JEOL Aluminium 32 mm
    100/pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 738,36 $
    738,36 $
    Each
       738,36 $
    Supplier Number: 75704