ContactUsLinkComponent
EKONO™ Hybridization Buffer, G-Biosciences
EKONO™ Hybridization Buffer, G-Biosciences
Catalog # CA82021-338
Supplier:  G-Biosciences
CAS Number:  
undefined
EKONO™ Hybridization Buffer, G-Biosciences
Catalog # CA82021-338
Supplier:  G-Biosciences
Supplier Number:  786-160
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EKONO* Hybridiztion Buffer
  • Size:
    1 L
  • Cat. no.:
    CA82021-338
Specifications
About this item
A high performance hybridization buffer system providing researchers with unprecedented simplicity, reliability, accuracy, and cost saving for hybridization
Protocol involves incubation of blots with denatured probes in EKONO™ hybridization buffer followed by washing of the blot to remove excess unhybridized probes. Easy-to-use EKONO™ provides high resolution and sensitivity.
Suitable for Southern/Northern or Southwestern hybridization analysis, hybridization with oligonucleotides or other types of probes, and for hybridization screening of gene libraries.
EKONO™ hybridization buffer is suitable for all types of probes