Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Marketsource
Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Long handles ideal for acquiring samples from bottles, jars, and more.
  • Length 17.8 cm
  • Non-sterile
  • Translucent
  • Available in 3 bowl sizes
Long-handled spoons are ideal for acquiring samples from bottles, jars, and other confined areas and helping to avoid personal contact with sample materials. A flat edge on the rim of each bowl allows fast levelling to collect and measure small amounts with accuracy. Each spoon is 17.8 cm (7") overall length. Non-sterile and translucent in color.
Product Family Options
  • Désignation
    Capacité
  • Sample Spoon
    1 tsp.
    Each
    Each - $75.93
    $75.93
    Pack of 10
    Pack of 10 - $46.61
    $46.61
    Each
       $75.93
    Pack of 10
       $46.61
     : 72133
    Marketsource
    Marketsource
  • Sample Spoon
    ¼ tsp.
    Each
    Each - $54.35
    $54.35
    Each
       $54.35
     : 72131
    Marketsource
    Marketsource
  • Sample Spoon
    ½ tsp.
    Each
    Each - $72.96
    $72.96
    Pack of 10
    Pack of 10 - $46.39
    $46.39
    Each
       $72.96
    Pack of 10
       $46.39
     : 72132
    Marketsource
    Marketsource