Polytron® Generator PT-DA 20/2FEC-E193, Kinematica
PT-DA 25 / 2FEC-E192, Fine Slot, 100 - 2500 ml, 20 mm, 20×192 mm, PT 2500 E
Catalog # 97057-708
Supplier: Kinematica
Supplier #: 11030265
Jump to:
Product Details & Documents
F style generator for creating emulsions
- Easy clean design for quick disassembly and cleaning
- Quick connect coupling
- F syle with finely slotted rotor generates maximum shear forces
- Stainless steel (316L)
This generator is designed especailly for creating stable emulsions. With the 30000 rpm motor tips speeds of 27 m/sec can be achieved.
Rotor designed for maximum shear forces.
Spécifications
- Ø:20 mm
- Désignation:PT-DA 25 / 2FEC-E192, Fine Slot
- Diameter ×L:20×192 mm
- For Use With:PT 2500 E
- Sample Volume:100 - 2500 ml