Order Entry
Canada
ContactUsLinkComponent
(Arg)9, TAMRA (5-Carboxytetramethylrhodamin), AnaSpec
Catalogue # 103006-414
Fournisseur:  Anaspec Inc
Numéro CAS:  
(Arg)9, TAMRA (5-Carboxytetramethylrhodamin), AnaSpec
Catalogue # 103006-414
Fournisseur:  Anaspec Inc
Supplier Number:  AS-61208
Numéro CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Spécifications

  • Conjugation:
    TAMRA (5-Carboxytetramethylrhodamin)
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Cell permeable Peptide|Arginine repeat|CPP
  • Protein/peptide name:
    (Arg)9
  • Purity:
    95%
  • Molecular weight:
    1836.2 Da
  • Sequence:
    TAMRA-RRRRRRRRR
  • Cat. no.:
    103006-414
  • Supplier No.:
    AS-61208

Spécifications

À propos de cet article

(Arg)9 is a cell-permeable peptide used for drug delivery.It can traverse the plasma membrane of eukaryotic cells. This is a fluorescent (TAMRA)-labeled cell permeable peptide, Abs/Em=541/568.
Sequence:TAMRA-RRRRRRRRR
MW:1836.2 Da
% peak area by HPLC:95
Storage condition:-20° C