Order Entry
Canada
ContactUsLinkComponent
[Lys(Ac) 5/8/12/16]-Histone H4 (1-25)-GSGSK,Biotin
Catalogue # 103009-196
Fournisseur:  Anaspec Inc
Numéro CAS:  
[Lys(Ac) 5/8/12/16]-Histone H4 (1-25)-GSGSK,Biotin
Catalogue # 103009-196
Fournisseur:  Anaspec Inc
Supplier Number:  AS-65248-1
Numéro CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Spécifications

  • Conjugation:
    Biotin
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Histone H4 Peptide
  • Protein/peptide name:
    [Lys(Ac) 5/8/12/16]-Histone H4 (1-25)-GSGSK
  • Purity:
    95%
  • Molecular weight:
    3400.9 Da
  • Sequence:
    SGRG-K(Ac)-GG-K(Ac)-GLG-K(Ac)-GGA-K(Ac)-RHRKVLRDN-GSGSK(Biotin)
  • Cat. no.:
    103009-196
  • Supplier No.:
    AS-65248-1

Spécifications

À propos de cet article

This peptide is histone H4 (1-25) with acetylation at Lys5, Lys8, Lys12, and Lys16, followed by a C-terminal GSGS linker and a biotinylated lysine. Acetylation at Lys5, Lys8, and Lys12 is carried out by p300 protein while acetylation at Lys16 is regulated by several histone acetyltransferases. These lysine residues have been shown to have important functions in transcriptional activation, replication, and nuclear division. Acetylation of lysine residues promotes binding of transcription factors to histone H4. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:SGRG-K(Ac)-GG-K(Ac)-GLG-K(Ac)-GGA-K(Ac)-RHRKVLRDN-GSGSK(Biotin)
MW:3400.9 Da
% peak area by HPLC:95
Storage condition:-20° C