Order Entry
Canada
ContactUsLinkComponent
 
[Lys(Me2)4]-Histone H3 (1-21)
  103007-648
 :  
[Lys(Me2)4]-Histone H3 (1-21)
  103007-648
 :  AS-63677
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Histone H3 Peptide|H3K4
  • Protein/peptide name:
    [Lys(Me2)4]-Histone H3 (1-21)
  • Purity:
    95%
  • Molecular weight:
    2282.6 Da
  • Sequence:
    ART-K(Me2)-QTARKSTGGKAPRKQLA
  • Cat. no.:
    103007-648
  • Supplier No.:
    AS-63677

 

 

This is a dimethylated lysine at position 4 of the 1-21 amino acid histone H3 peptide. This form of histone methylation is characterized as being enriched around transcription start sites (TSS). It has been observed that a subgroup of genes linked to T cell functions showed high levels of H3K4me2 within their gene body. While enrichment for H3K4me2 within the gene body is a characteristic trait of expressed genes in yeast, in contrast, H3K4me2 has been shown to be predominantly enriched around the TSS in mammals.
Sequence:ART-K(Me2)-QTARKSTGGKAPRKQLA
MW:2282.6 Da
% peak area by HPLC:95
Storage condition:-20° C