Order Entry
Canada
ContactUsLinkComponent
Hen HEL (46-61)
Catalogue # 103003-186
Fournisseur:  Anaspec Inc
Numéro CAS:  
Hen HEL (46-61)
Catalogue # 103003-186
Fournisseur:  Anaspec Inc
Supplier Number:  AS-60504-1
Numéro CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Spécifications

  • Conjugation:
    Unconjugated
  • Species:
    Hen
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    hen egg lysozyme Peptide fragment|hen egg lysozyme 46-61
  • Protein/peptide name:
    HEL (46-61)
  • Purity:
    95%
  • Molecular weight:
    1753.9 Da
  • Sequence:
    NTDGSTDYGILQINSR
  • Cat. no.:
    103003-186
  • Supplier No.:
    AS-60504-1

Spécifications

À propos de cet article

This peptide is amino acids 46 to 61 fragment of the hen egg lysozyme (HEL). Lysozyme is anti-bacterial enzyme found in high concentration in the egg white. HEL was used in MHC related studies, and tested as a part of antimicrobial vaccines.
Sequence:NTDGSTDYGILQINSR
MW:1753.9 Da
% peak area by HPLC:95
Storage condition:-20° C