To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:In-Gel Tryptic Digestion Kit
- Cat. no.:CAPI89871
- No. of Reactions:150
- Supplier No.:89871
Spécifications
À propos de cet article
The Pierce™ In-Gel Tryptic Digestion Kit provides a complete set of reagents for digesting and recovering peptide fragments from stained protein bands excised from SDS-PAGE (polyacrylamide gel plugs).
- Contains highly purified and modified MS-grade trypsin that shows no chymotryptic activity and minimal autolytic activity
Includes all necessary reagents for destaining Coomassie®- or fluorescent dye-stained proteins, reduction and alkylation of cystines, and tryptic digestion. Procedure and reagents produce reliable digestions and data generation using a wide range of conditions without optimization.