Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Bradykinin antagonist|Bradykinin HOE I40
- Protein/peptide name:HOE I40
- Purity:95%
- Molecular weight:1304.5 Da
- Sequence:rRP-(Hyp)-G-(Thi)-S-(D-Tic)-(Oic)-R
- Cat. no.:102996-350
- Supplier No.:AS-22968
Hoe 140 is a stable B2 bradykinin antagonist. Based on its high potency and good tolerability, Hoe 140 is used to evaluate the role of bradykinin in human diseases.
Sequence:rRP-(Hyp)-G-(Thi)-S-(D-Tic)-(Oic)-R
MW:1304.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C