Order Entry
Canada
ContactUsLinkComponent
Human Thrombin Receptor Agonist
Catalogue # 103003-346
Fournisseur:  Anaspec Inc
Numéro CAS:  
Human Thrombin Receptor Agonist
Catalogue # 103003-346
Fournisseur:  Anaspec Inc
Supplier Number:  AS-60680
Numéro CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Spécifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    5 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Thrombin agonist amide|Agonist receptor thrombin|Thrombin agonist receptor
  • Protein/peptide name:
    Thrombin Receptor Agonist
  • Purity:
    95%
  • Molecular weight:
    634.1 Da
  • Sequence:
    SFLLR - NH2
  • Cat. no.:
    103003-346
  • Supplier No.:
    AS-60680

Spécifications

À propos de cet article

A protease-activated receptor (PAR-1) agonist peptide (P5-NH2) identified as the minimal structural motif required for retaining the full agonist activity.
Sequence:SFLLR - NH2
MW:634.1 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C