This sequence acts as a tethered peptide ligand. Free SFLLRN activates PAR1 independent of receptor cleavage and has been used to probe PAR1 function in various cells and tissues. This peptide is also known to be capable of activating PAR2.
Sequence:SFLLRN
MW:748.9 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Human
- Storage conditions:–20 °C
- Protein synonyms:TRAP|TRAP 6|Thrombin receptor activator
- Protein/peptide name:Thrombin Receptor Activator for Peptide 6
- Purity:95%
- Molecular weight:748.9 Da
- Sequence:SFLLRN