Plexiglas Microscope Slides, Electron Microscopy Sciences
Plexiglas Microscope Slides, Electron Microscopy Sciences
Plexiglass slides, 1.0 mm, 75×25 mm (3×1")
  103302-388
Supplier #:  71891-10
Plexiglas Microscope Slides, Electron Microscopy Sciences
  103302-388
Plexiglas Microscope Slides, Electron Microscopy Sciences
Plexiglass slides, 1.0 mm, 75×25 mm (3×1")
$96.79
Each
A Fool-Proof method for the mounting of serial sections on Single Hole Grids.
They prevent wrinkling and section loss while mounting sections on the grid.
  • Désignation:
    Plexiglass slides
  • L×l:
    75×25 mm (3×1")
  • Épaisseur:
    1.0 mm