À propos de cet article
Residual Oil Remover is designed to emulsify and remove microscopic oil residues a by-product of environmental pollution from the surface and pores of glass and plastic.
- Increases brightness and clarity
- Safe and versatile for delicate surfaces
- Available in either a dropper bottle or a Pump style bottle
ROR has been tested and used on everything from multicoated lenses to beryllium surfaced contacts and has proven time and again to be the finest and safest optical cleaning treatment.
ROR is used today by the military, NASA, astronomers, scientists, hospitals, laboratories and universities.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.