ContactUsLinkComponent
Pierce I-PER™, Insect Cell Protein Extraction Reagent, Thermo Scientific
Catalogue # CAPI89802
Fournisseur:  Thermo Scientific
Numéro CAS:  
undefined
Pierce I-PER™, Insect Cell Protein Extraction Reagent, Thermo Scientific
Catalogue # CAPI89802
Fournisseur:  Thermo Scientific
Supplier Number:  89802
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    I-PER™ Insect Cell Protein Extraction Reagents, for 5 – 20 Million Cultured Cells per mL of Reagent
  • Size:
    250 mL
  • Cat. no.:
    CAPI89802
  • Application:
    Protein Assays, Western Blotting, 6xHis-Tagged Protein Purifications
  • Supplier No.:
    89802
Spécifications
À propos de cet article
I-PER™ Insect Cell Protein Extraction Reagent enables gentle and effective extraction of soluble protein from baculoviral-infected insect cells grown in suspension or monolayer culture.
  • Gentle extraction - optimized, mild non-ionic detergent provides maximum extraction of soluble proteins from insect cells
  • Effective - provides better protein extraction than sonication
  • Compatible - downstream compatibility with Western blotting, 6xHis-tagged protein purification, protein assays and ion-exchange chromatography
  • Flexible - useful for protein extraction from suspended or adherent cultured insect cells
The baculoviral insect cell expression system is an efficient and popular system for production of recombinant (eukaryotic) proteins in cell culture. Proteins expressed in baculoviral systems can be used for structural analyses, biochemical assays and a variety of other applications. I-PER Reagent maintains functionality of extracted proteins and is directly compatible with downstream applications such as protein assays, Western blotting and 6xHis-tagged protein purification.