ImmunoHistoZyme™, Ready-to-Use, Electron Microscopy Sciences
ImmunoHistoZyme™, Ready-to-Use, Electron Microscopy Sciences
ImmunoHistoZyme™, Ready-to-Use, Electron Microscopy Sciences
This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.
In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC
Preparation of Working Solution: Dilute this 10X buffer as needed (e.g. 90 mL of deionized or distilled water+10 mL of this buffer) mix well, 1X buffer can be stored at 2 to 8 °C.
The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.
Product Family Options
  • Désignation
    Taille
  • ImmunoHistoZyme™, Ready-to-Use
    15 mL
    Each (15ml)
    Each (15ml) - $202.47
    $202.47
    Each (15ml)
       $202.47
     : 64142-13
  • ImmunoHistoZyme™, Ready-to-Use
    100 mL
    Each (100ml)
    Each (100ml) - $383.90
    $383.90
    Each (100ml)
       $383.90
     : 64142-14