Orbital Specimen Holder, Electron Microscopy Sciences | Avantor
Orbital Specimen Holder, Electron Microscopy Sciences
Orbital Specimen Holder, Electron Microscopy Sciences
Certificates
Orbital Specimen Holder, Electron Microscopy Sciences
Orbital Specimen Holder, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Unique orbital specimen holder for a spin and raise SEM images to a whole new level.
  • Rotate 360° around the axis perpendicular to the electron beam
  • Position the sample with turn of a screw
  • Flexibile and superior control
The pot is coated to reduce noise, producing a black background behind the subject so every detail stands out, allowing the capture of stunning, clear images.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Orbital Specimen Holder Pin Mount
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $632.22
    $632.22
    Each
       $632.22
    Supplier Number: 75911
  • Orbital Specimen Holder M4 Mount
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $617.95
    $617.95
    Each
       $617.95
    Supplier Number: 75912