Tweezers WA1 Handle with 55-STD Tips, Electron Microscopy Sciences | Avantor
Tweezers WA1 Handle with 55-STD Tips, Electron Microscopy Sciences
Tweezers WA1 Handle with 55-STD Tips, Electron Microscopy Sciences
Certificates
Tweezers WA1 Handle with 55-STD Tips, Electron Microscopy Sciences
Tweezers WA1 Handle with 55-STD Tips, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These tweezers have an anti-magnetic WA1 handle with replaceable 55-STD tips.
  • Hand made of the highest quality steels and alloys
  • Tips attach with screws
  • High precision and perfectly finished to suit the application
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as inox.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Style
    Tip Thickness
    Tip Width
    Availability
    Price
  • Handle with 55-STD Replaceable Tips
    Antimagnetic
    115 mm
    Grey STD
    0.7 mm
    2 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $102.59
    $102.59
    Each
       $102.59
    Supplier Number: 10004-S55-SM
  • 55-STD Replaceable Tips
    -
    Grey STD
    0.7 mm
    2 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $20.92
    $20.92
    Each
       $20.92
    Supplier Number: TIP-55-STD