SEM Bulk Specimen Holder, Electron Microscopy Sciences
SEM Bulk Specimen Holder, Electron Microscopy Sciences
Marketsource
SEM Bulk Specimen Holder, Electron Microscopy Sciences
Marketsource
Certificates
SEM Bulk Specimen Holder, Electron Microscopy Sciences
SEM Bulk Specimen Holder, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For clamping large irregular, bulky samples to view in a scanning electron microscope.
  • Made of machined aluminum with stainless steel allen set screws
  • Pin Diameter: 3.2 mm
Delivery information: Allen wrench included.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Bulk Specimen Holder
    24.5 x 15.9 x 9.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $247.79
    $247.79
    Each
       $247.79
    Supplier Number: 75951-06
  • Large Bulk Specimen Holder
    32 x 32 x 13 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $305.40
    $305.40
    Each
       $305.40
    Supplier Number: 75951-07
    Spécifications:
    Marketsource
    Marketsource
  • X-Large Bulk Specimen Holder
    40 x 52 x 13 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $373.27
    $373.27
    Each
       $373.27
    Supplier Number: 75951-08
  • XX-Large Bulk Specimen Holder
    48 x 80 x 13 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $421.58
    $421.58
    Each
       $421.58
    Supplier Number: 75951-09
    Spécifications:
    Marketsource
    Marketsource