3 D Stereo Glasses, Electron Microscopy Sciences
À propos de cet article
White cardboard frames with temples for Red-Cyan or Red-Blue 3D anaglyph needs.
With red and cyan filters or Red and Blue filters, the glasses can be used for either red left or red right depending on which way you fold the temples.
Two different fold-scores allows to fit the glasses to smaller or larger viewer's heads.
2 Options Available Below
Product Details & Documents
White cardboard frames with temples for Red-Cyan or Red-Blue 3D anaglyph needs.
With red and cyan filters or Red and Blue filters, the glasses can be used for either red left or red right depending on which way you fold the temples.
Two different fold-scores allows to fit the glasses to smaller or larger viewer's heads.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 68139-10Spécifications:MarketsourceMarketsource
- 3D Glasses, Red-BlueEach$14.57Pack of 10$10.97Supplier Number: 68139-20Spécifications:MarketsourceMarketsource