EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
Certificates
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.
In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC
Preparation of Working Solution: Dilute this 10X buffer as needed (90 mL of deionized or distilled water+10 mL of this buffer) mix well, 1X buffer can be stored at 2 to 8 °C.
The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.
Product Family Options
Product Information
  • Désignation
    Taille
    pH
    Availability
    Price
  • EDTA Buffer
    100 mL
    8.5
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $191.72
    $191.72
    Each (100ml)
       $191.72
    Supplier Number: 64142-09
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • EDTA Buffer
    1000 mL
    8.5
    Log in for availability
    Log in for availability
    Each (1,000ml)
    Each (1,000ml) - $390.61
    $390.61
    Each (1,000ml)
       $390.61
    Supplier Number: 64142-10
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.