Saws, Electron Microscopy Sciences | Avantor
Saws, Electron Microscopy Sciences
Saws, Electron Microscopy Sciences
Certificates
Saws, Electron Microscopy Sciences
Saws, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Three styles of manual bone saws are available.
  • Broad bladed Satterlee Saw
  • Thinner rounded top Charriere Saw
  • Solid handle Langenbech Metacarpal Saw
The broad bladed Satterlee Saw (A), the thinner rounded tip Charriere Saw (B), and the solid handle Langenbech Metacarpal Saw (C). Each saw has serrated teeth.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Satterlee Saw Replacement Blade
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $192.42
    $192.42
    Each
       $192.42
    Supplier Number: 70566-32
  • Charriere Saw, 267 mm Long
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $266.52
    $266.52
    Each
       $266.52
    Supplier Number: 70566-30
  • Satterlee Saw, 292 mm Long
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $352.76
    $352.76
    Each
       $352.76
    Supplier Number: 70566-31