Section Block, Electron Microscopy Sciences
Section Block, Electron Microscopy Sciences
Section Block, Electron Microscopy Sciences
Blocking solutions based on chemically modified and fragmented ultra-pure casein. Effectively reduces unwanted binding of primary antibody and conjugates use to charged surface of the slide and tissue section.
Greatly reduces non-specific binding while preserving the specific reaction, by saturating potential non-specific protein-protein interactions.
Recommended for research and diagnostic pathology, especially for retrieved sections and polyclonal antibodies.
Product Family Options
  • Désignation
    Taille
  • Section Block
    50 mL
    Each (50ml)
    Each (50ml) - $361.16
    $361.16
    Kit of 50
    Kit of 50 - $361.16
    $361.16
    Each (50ml)
       $361.16
    Kit of 50
       $361.16
     : 62710
  • Section Block
    125 mL
    Each (125ml)
    Each (125ml) - $777.69
    $777.69
    Each (125ml)
       $777.69
     : 62711
  • Section Block
    500 mL
    Each (500ml)
    Each (500ml) - $2,828.21
    $2,828.21
    Each (500ml)
       $2,828.21
     : 62712