Athene Square Grids, Electron Microscopy Sciences
À propos de cet article
Highest quality square mesh transmission electron microscope grids.
- Copper and nickel grids
- 50, 150, 200, 300, and 400 mesh grids
- Good handling characteristics
Individually inspected before being packed.
7 Options Available Below
Product Details & Documents
Highest quality square mesh transmission electron microscope grids.
- Copper and nickel grids
- 50, 150, 200, 300, and 400 mesh grids
- Good handling characteristics
Individually inspected before being packed.
Product Family Options
Informations sur le produit
- DésignationMatièreØMeshAvailabilityPrice
- Supplier Number: A50-CU
- Supplier Number: A150-CU
- Square GridsCopper3.05 mm200Each$118.82Pack of 100$91.23Supplier Number: A200-CUSpécifications:MarketsourceMarketsource
- Square GridsNickel3.05 mm200Each$138.36Pack of 100$112.02Supplier Number: A200-NISpécifications:MarketsourceMarketsource
- Supplier Number: A300-NI
- Square GridsCopper3.05 mm300Each$103.73Pack of 100$81.21Supplier Number: A300-CUSpécifications:MarketsourceMarketsource
- Supplier Number: A400-CU