Bacti-Cinerator IV, Electron Microscopy Sciences | Avantor
Bacti-Cinerator IV, Electron Microscopy Sciences
Replacement Bacti-Cinerator III Heat Element, 115 V
Catalog # 101412-628
Fournisseur #:  62420-51
Bacti-Cinerator IV, Electron Microscopy Sciences
Catalog # 101412-628
Bacti-Cinerator IV, Electron Microscopy Sciences
Bacti-Cinerator IV, Electron Microscopy Sciences
Replacement Bacti-Cinerator III Heat Element, 115 V
Price
$910.13
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Baci-Cinerator IV sterilizes by infrared heat (Heater Element contains no asbestos) - organic material is incinerated deep inside the ceramic tunnel. To use, simply insert loop or needle into the tunnel. Complete sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500 °F.
  • Prevents infectious spatter and/or cross contamination
  • No open flame -no hazardous gas
  • No need to wait for the loops or needles to turn red
  • Does not require oxygen so it can be used in anaerobic chambers
  • UL approved
Spécifications
  • Désignation:
    Replacement Bacti-Cinerator III Heat Element
  • Electrical:
    115 V