Distillation Rods, Büchi | Avantor
Distillation Rods, Büchi
Distillation Rods, Set of 10
Catalog # 76169-032
Supplier:   Buchi
Fournisseur #:  043087
Distillation Rods, Büchi
Catalog # 76169-032
Supplier:  Buchi
Distillation Rods, Büchi
Distillation Rods, Büchi
Distillation Rods, Set of 10
Price
$3,020.38
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Reusable glass rods designed specifically to aid in digestion of samples for Kjeldahl determination.
  • Prevent bumping
  • Minimize foaming
  • Designed not to interfere with aspiration of automated systems
Aids in minimizing foaming of high surface tension samples to keep entire sample contents within the reflux zone. Helps to equilibrate heat evenly throughout the samples.
Spécifications
  • Désignation:
    Distillation Rods, Set of 10