Float Racks for Microtubes, Globe Scientific | Avantor
Float Racks for Microtubes, Globe Scientific
Supplier:   Globe Scientific
Float Racks for Microtubes, Globe Scientific
Supplier:  Globe Scientific
Certificates
Float Racks for Microtubes, Globe Scientific
Float Racks for Microtubes, Globe Scientific
2 Options Available Below
Product Details & Documents
Autoclavable polypropylene construction for strength, buoyancy, and temperature resistance.
  • Racks float with full load of filled tubes
  • Molded handle in center of rack for comfort and support during transport
  • Legs allow rack to stand upright for use on benchtop
  • Use in ice, water baths, or on the benchtop
  • 65 mm round rack fits most 400 mL beakers
Commonly found in cloning, microbiology, and immunology labs.
Product Family Options
Square
Informations sur le produit
  • Désignation
    Color
    Nb de places
    Availability
    Price
  • 95 x 95 x 80 mm (3.75 x 3.75 x 3")
    White
    16
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - 69,58 $
    69,58 $
    Prix promotionnel pour les nouveaux clients
    Pack of 4
       69,58 $
    Supplier Number: 456395
Round
Informations sur le produit
  • Désignation
    Color
    Nb de places
    Availability
    Price
  • 65 mm (2.5 dia)
    White
    8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - 49,73 $
    49,73 $
    Prix promotionnel pour les nouveaux clients
    Pack of 4
       49,73 $
    Supplier Number: 456398