ContactUsLinkComponent
Methionine Sulfoxide Immunoblotting Kit, Cayman Chemical
  75816-974
 :  
undefined
Methionine Sulfoxide Immunoblotting Kit, Cayman Chemical
  75816-974
 :  600160-1
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Brand name:
    CAYM
  • Cat. no.:
    75816-974
  • Contents:
    Kit contains: Antibody, Positive Control, and Blocking Reagent
  • Supplier No.:
    600160-1
For the immunochemical detection of proteins containing MetO residues by western blotting.
  • Detect proteins containing MetO residues by Western blot
  • Positive control included
Protein MetO is a reversible oxidative modification that occurs by exposure of protein(s) methionine residues to reactive oxygen species (ROS). Methionine oxidation can alter the function(s) of the modified proteins and if not reversed by MetO reductases can be further oxidized to methionine sulfone, an irreversible modification. Cayman’s MetO Immunoblotting Kit contains reagents needed for the immunochemical detection of proteins containing MetO residues by western blotting. MetO-containing samples of interest include those from cell or tissue lysates as well as semi-pure or purified proteins.
 : Positive control included.