Prostaglandin D₂-MOX ELISA Kit, Cayman Chemical
Competitive ELISA for quantitation of PGD2 in biological samples
- Treatment of samples with MOX hydrochloride converts unstable PGD2 to stable PGD2-MOX
- Highly sensitive
- Measure PGD2-MOX levels down to 3.1 pg/ml
- Incubation: 18 hours | Development: 90-120 minutes | Read: Colorimetric at 405-420 nm
- Midpoint: 50% B/B0: 10-35 pg/ml
This PGD2-MOX ELISA is based on the conversion of PGD2 to a stable MOX derivative. Treatment of the sample with MOX HCl converts PGD2 into PGD2-MOX, preventing its further chemical degradation. The antiserum used in the assay was developed using conjugates of this derivative and is very specific for PGD2-MOX.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Assay Type:Competitive
- Detection range:2.0 – 250 pg/ml
- Sensitivity:80% B/B0: 3 pg/mL
Frequently Bought Together