ContactUsLinkComponent
Protein Carbonyl Colorimetric Assay Kit, Cayman Chemical Company
  75816-674
 :  
undefined
Protein Carbonyl Colorimetric Assay Kit, Cayman Chemical Company
  75816-674
 :  10005020-96
 :  
  • Brand name:
    CAYM
  • Cat. no.:
    75816-674
  • Contents:
    Kit contains: Hydrochloric Acid, DNPH, Solution, Guanidine Hydrochloride, Ethanol, Ethyl Acetate, and Plate
  • Supplier No.:
    10005020-96
Convenient colorimetric assay for measurement of oxidized proteins.
  • Measure protein carbonyl content in plasma, serum, urine, tissue homogenates, and cell lysates
  • Assay 24 samples in duplicate
  • Plate-based colorimetric measurement (360 – 385 nm)
The carbonyls of protein samples are derivatized using 2,4-dinitrophenylhydrazine. Formation of a Schiff base produces the corresponding hydrazone which can be analyzed spectrophotometrically at 360 – 385 nm. This assay can be used to measure oxidized protein in plasma, serum, cell lysates, and tissue homogenates.