Gamma Wipe® - VP Sterile Sealed Edge Cleanroom Laundered 100% Polyester Knit Wipes, Berkshire
76170-274
- Sterility:Sterile
- Description:Gamma Wipe® - VP, Ultrasonically Sealed
- Material:Knitted Polyester
- Cat. no.:76170-274
- Packaging:20/Pack, Stacked
- W×L:30.5×30.5 cm (12×12")
Ultrasonically sealed edge for reduced fiber contamination. Gamma irradiated and sterile validated to a 10⁻⁶ Sterility Assurance Level per AAMI guidelines.
- Light weight
- High absorbency material design reduces actual cost in use compared to heavier weight products
The Gamma Wipe® - VP offers critically low particle, fiber, ion, and extracble levels as well as abrasion resistance and absorbency.
Chemically compatible with common cleaning and disinfecting solutions including isopropyl alcohol, phenols, bleaches, and quarternary ammonium compounds. Complies with RoHS and REACH requirements. Designed for cleaning sterile surfaces and equipment during aseptic processing. Excellent for cleaning sterile laminar air flow units, barrier isolators, and bio-containment units.
Radiation dosage and lot information provided.
: ISO Class 3–5. Cleanroom Class 1–100. EU Grade A/B/C/D. USP 797. Laundered and packaged in ISO Class 4 cleanroom with a lot number on each pack. Meets cGMP requirements for traceability.