Drug Testing Model Set
Drug Testing Model Set
Catalog # 470232-070
Supplier: Mega Molecules
Supplier #: FC125
Jump to:
Product Details & Documents
Hands on representation of drug testing models.
- Includes 1 activity
- Perfect for high school students
In this activity, students act as members of a hospital administration who are responsible for developing an effective substance-abuse prevention and drug testing program. Students will explore the chemical and physical properties of five illicit drugs and build a model of each drug. Students will analyze the results of urine test data and evaluate drug use among hospital applicants and employees.
Specifications
- Désignation:Drug Testing Model Set