À propos de cet article
MPure Viral/Pathogen Nucleic Acids Extraction Kit B is used with the MPure 12 instrument for extraction of viral and bacterial DNA/RNA from Viral, bacterial and swab samples (cell-rich samples).
- Extraction of viral and bacterial DNA/RNA
- Completely automated purification protocol
- Use with MPure-12 System
- Can be run in as little as 55 minutes
- Extracted Nucleic Acids can be used in downstream applications
MPure™ Viral/Pathogen Nucleic Acids Extraction Kit B is used with MPure-12™ instrument for extraction of viral and bacterial DNA/RNA from bacterial culture, swab samples and environmental samples. Starting samples include clinical specimens such as Sputum, Bronchoalveolar lavage, cell-free body fluids, urine, respiratory specimens, bacterial culture either solid or in suspension and environmental samples such as soil, water and feces. The kit contains everything needed for sample preparation. However, for tough samples (soil or feces), sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten procesing times.
Sample Type: Swab, Bacterial, Viral
Sample Volume: Bacteria Pellet: 100-200 µl /Up to 109 bacteria (about OD600 = 3); Bacterial Colony: 100-200 µl /1-3 colony; Swab samples: 100-200 µl liquid transport media
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Viral RNABacterial RNAViral DNABacterial DNA
- Sample type:SwabsBiofluids (serum, plasma, saliva)
- Target molecule:Copurification of DNA & RNADNARNA
- Description:MPure™ Viral/Pathogen Nucleic Acids Extraction Kit B