About this item
The MPure Viral Nucleic Acid Extraction Kit is used with the MPure-12 instrument for the extraction of high quality viral DNA and RNA from serum, plasma or cell-free body fluids of 100 µL - 400 µL using proven magnetic particle separation technology.
- Proven for viral nucleic acids isolation of HIV, HCV, HBV, CMV and EBV, influenza viruses from plasma and serum and cell-free body fluids
- Ready-to-use prelled reagent cartridge
- Uniquely designed magnetic bead processing chamber providing the highest quality results and efficient removal of inhibitors
- High-quality Viral DNA/RNA for downstream application
Nucleic acids extracted can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, Southern Blot Analysis.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Viral RNAViral DNA
- Sample type:SwabsBiofluids (serum, plasma, saliva)
- Target molecule:Copurification of DNA & RNADNARNA
- Description:MPure™ Viral Nucleic Acid Extraction Kit