This US origin iron supplemented calf serum for use in cell culture media. Supplementing iron to formula fed calf serum can enhance serum transferrin levels and an alternative to FBS in promoting cell growth and its effectiveness.
- Sterile 0.2 µm filtered
- Maximize cell growth with Lipogro supplement
- Can produce large volume lot sizes which results in significantly better lot-to-lot consistency, so you are assured a high-performing, superior product
Raw materials are collected from calves ranging from 14 days and six months of age.
Manufactured at our state-of-the art facility which adheres to GMP standards and contains stringent process controls and validated CIP cleaning processes.
Bovine calf serum is carefully collected using well-established protocol and fully trained collection specialist.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.