Storage Vials, Quick/Snap, MicroSolv
Supplier: MicroSolv Technology Corp
Jump to:
À propos de cet article
These easy to use storage vials and caps use a push on technology for ease and comfort.
- Easy to handle
- Quick and easy
- No liners required in the cap
Use these vials to store samples that are not critical.
: Caps sold separately.
4 Options Available Below
Product Details & Documents
These easy to use storage vials and caps use a push on technology for ease and comfort.
- Easy to handle
- Quick and easy
- No liners required in the cap
Use these vials to store samples that are not critical.
: Caps sold separately.
Product Family Options
Informations sur le produit
- DésignationCapacitéDiameter ×HAvec bouchonTaille du bouchonAvailabilityPrice
- Supplier Number: 97010-CV05
- Supplier Number: 97010-CV10
- Supplier Number: 97010-CV15
- Supplier Number: 97010-CV25
Caps
Informations sur le produit
- Catalog #DésignationCap TypeCap ColorAvailabilityPrice
- Small caps, use with ND18 vials, 19.8 x 5.2 mmPE-push on capTransparentLog in pour la disponibilitéPack of 10018,72 $