Staticmaster® Electrostatic Discharge Workstation Kit, NRD
Electrostatic discharge workstation kit
Catalog # 75794-918
Supplier: NRD Staticmaster
Fournisseur #: 5021
Staticmaster® Electrostatic Discharge Workstation Kit, NRD
Catalog # 75794-918
Supplier: NRD Staticmaster
Jump to:
Product Details & Documents
The Staticmaster® 5021 ESD workstation kit provides an ESD safe work surface and grounding supplies for handling static sensitive products.
- Provides ESD safe work surface
- Includes grounded rubber table mat
The kit includes a 16" x 24" ESD rubber mat which has a dissipative top layer and a conductive bottom layer. The mat is grounded with a low profile common point ground and installed ground snap. A blue fabric wrist band and 6' coil cord grounds the operator.
Certifications: The kit meets or exceeds requirements of ANSI ESD S541-2003. RoHS Compliant.
Spécifications
- Désignation:Electrostatic discharge workstation kit