Staticmaster® Electrostatic Discharge Workstation Kit with Duostat Positioner, NRD
75794-894
- Description:ESD workstation kit with Duostat positioner
- Cat. no.:75794-894
- Supplier No.:SMF-KITALPHA
This Staticmaster® filter alpha kit includes the Duostat Positioner, allowing it to remove static charge on both sides of filters simultaneously before weighing
- Removes static charge on both sides of filters simultaneously
- Eliminates charge before weighing
Includes the ESD workstation kit, (3) 2U500 Ionizers (3617097), (1) BF3 EZ mount, and (1) 5060 grounding kit.