Staticmaster® Electrostatic Discharge Workstation Kit with Unistat Positioner, NRD
75794-892
- Description:ESD workstation kit with unistat positioner
- Cat. no.:75794-892
- Supplier No.:SMP-KITALPHA
This Staticmaster® powder alpha kit includes the Unistat Positioner, allowing it to bring ionization to the powder transfer process preventing fly away from static
- Brings ionization to the powder transfer process
- Prevents fly away from static
Includes ESD workstation kit, (2) 2U500 Ionizers (3617097), (1) BF EZ mount, and (1) 5060 grounding kit.